Learn More
Biomatik Corporation SPTLC2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508451
Encompass_Preferred
Sptlc2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SPTLC2. The Sptlc2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine Palmitoyltransferase 2 Protein (88-562Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SPTLC2. Accession Number: O15270. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8451 Supplier Biomatik Corporation Supplier No. CAC08868
Sptlc2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SPTLC2. The Sptlc2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine Palmitoyltransferase 2 Protein (88-562Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SPTLC2. Accession Number: O15270. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.