Learn More
Biomatik Corporation STAT3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08876
Encompass_Preferred
Stat3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to STAT3. The Stat3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Signal Transducer And Activator Of Transcription 3 Protein (50-240Aa). Applications: ELISA, IHC, IF, IP. Target Synonyms: STAT3. Accession Number: P40763. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8459