Learn More
Biomatik Corporation SUCNR1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC08883
Sucnr1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SUCNR1. The Sucnr1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Succinate Receptor 1 Protein (303-334Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SUCNR1. Accession Number: Q9BXA5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.