Learn More
Biomatik Corporation SYNJ2BP anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508472
Encompass_Preferred
Synj2Bp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SYNJ2BP. The Synj2Bp Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Synaptojanin-2-Binding Protein (1-117Aa). Applications: ELISA, WB, IHC, IP. Target Synonyms: SYNJ2BP. Accession Number: P57105. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8472 Supplier Biomatik Corporation Supplier No. CAC08889
Synj2Bp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SYNJ2BP. The Synj2Bp Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Synaptojanin-2-Binding Protein (1-117Aa). Applications: ELISA, WB, IHC, IP. Target Synonyms: SYNJ2BP. Accession Number: P57105. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.