Learn More
Biomatik Corporation SYNPR anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508473
Encompass_Preferred
Synpr Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SYNPR. The Synpr Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Synaptoporin Protein (199-265Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SYNPR. Accession Number: Q8TBG9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8473 Supplier Biomatik Corporation Supplier No. CAC08890
Synpr Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SYNPR. The Synpr Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Synaptoporin Protein (199-265Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SYNPR. Accession Number: Q8TBG9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.