Learn More
Biomatik Corporation TMEM165 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08916
Encompass_Preferred
Tmem165 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TMEM165. The Tmem165 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transmembrane Protein 165 Protein (173-228Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TMEM165. Accession Number: Q9HC07. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8499