Learn More
Biomatik Corporation TMEM176B anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08917
Encompass_Preferred
Tmem176B Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TMEM176B. The Tmem176B Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transmembrane Protein 176B Protein (1-64Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TMEM176B. Accession Number: Q3YBM2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8500