Learn More
Biomatik Corporation TMEM192 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08920
Encompass_Preferred
Tmem192 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TMEM192. The Tmem192 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transmembrane Protein 192 Protein (193-271Aa). Applications: ELISA, WB, IHC, IF, IP. Target Synonyms: TMEM192. Accession Number: Q8IY95. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8503