Learn More
Biomatik Corporation EMC6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08932
Encompass_Preferred
The EMC6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ER membrane protein complex subunit 6 protein (2-47AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: ER membrane protein complex subunit 6, EMC6, TMEM93, Transmembrane protein 93. UniProt: Q9BV81. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8515