Learn More
Biomatik Corporation Tnfsf11 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08937
Encompass_Preferred
Tnfsf11 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Tnfsf11. The Tnfsf11 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Tumor Necrosis Factor Ligand Superfamily Member 11 Protein (73-306Aa). Applications: ELISA, IF. Target Synonyms: Tnfsf11. Accession Number: O35235. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8520