Learn More
Biomatik Corporation Tpsb2 anti-Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08943
Encompass_Preferred
Tpsb2 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Tpsb2. The Tpsb2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Tryptase Beta-2 Protein (32-276Aa). Applications: ELISA, WB. Target Synonyms: Tpsb2. Accession Number: P21845. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8526