Learn More
Biomatik Corporation TRIM37 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08948
Encompass_Preferred
Trim37 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRIM37. The Trim37 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human E3 Ubiquitin-Protein Ligase Trim37 Protein (301-600Aa). Applications: ELISA, IHC, IF. Target Synonyms: TRIM37. Accession Number: O94972. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8531