Learn More
Biomatik Corporation TSPAN5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08955
Encompass_Preferred
Tspan5 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TSPAN5. The Tspan5 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tetraspanin-5 Protein (114-232Aa). Applications: ELISA, WB, IF. Target Synonyms: TSPAN5. Accession Number: P62079. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8538