Learn More
Biomatik Corporation VPS4B anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08972
Encompass_Preferred
Vps4B Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to VPS4B. The Vps4B Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Vacuolar Protein Sorting-Associated Protein 4B Protein (1-117Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: VPS4B. Accession Number: O75351. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8555