Learn More
Biomatik Corporation YKT6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08978
Encompass_Preferred
Ykt6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to YKT6. The Ykt6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Synaptobrevin Homolog Ykt6 Protein (10-195Aa). Applications: ELISA, WB, IHC. Target Synonyms: YKT6. Accession Number: O15498. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8561