Learn More
Biomatik Corporation ZNF133 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508564
Encompass_Preferred
Znf133 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZNF133. The Znf133 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Zinc Finger Protein 133 Protein (1-210Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: ZNF133. Accession Number: P52736. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8564 Supplier Biomatik Corporation Supplier No. CAC08981
Znf133 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZNF133. The Znf133 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Zinc Finger Protein 133 Protein (1-210Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: ZNF133. Accession Number: P52736. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.