Learn More
Biomatik Corporation ZNF695 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09005
Encompass_Preferred
Znf695 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZNF695. The Znf695 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Zinc Finger Protein 695 Protein (1-88Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: ZNF695. Accession Number: Q8IW36. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8588