Learn More
Biomatik Corporation EIF3I anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09020
Encompass_Preferred
The EIF3I Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Eukaryotic translation initiation factor 3 subunit I protein (1-323AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:2000-1:10000, IF:1:50-1:500. UniProt: Q13347. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8603