Learn More
Biomatik Corporation BCAP31 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09027
Encompass_Preferred
The BCAP31 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human B-cell receptor-associated protein 31 protein (2-243AA). Applications: ELISA, WB, IHC, IF, IP. Recommended dilution: WB:1:1000-1:5000, IHC:1:1000-1:2000, IF:1:50-1:500, IP:1:200-1:2000. UniProt: P51572. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8610