Learn More
Biomatik Corporation C1D anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09029
Encompass_Preferred
The C1D Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Nuclear nucleic acid-binding protein C1D protein (1-135AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:100-1:500, IF:1:50-1:500. Alternative Names: hC1D, C1D. UniProt: Q13901. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8612