Learn More
Biomatik Corporation RAC1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508626
Encompass_Preferred
Rac1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RAC1. The Rac1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related C3 Botulinum Toxin Substrate 1 Protein (11-185Aa). Applications: ELISA, IHC, IF. Target Synonyms: RAC1. Accession Number: P63000. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8626 Supplier Biomatik Corporation Supplier No. CAC09043
Rac1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RAC1. The Rac1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related C3 Botulinum Toxin Substrate 1 Protein (11-185Aa). Applications: ELISA, IHC, IF. Target Synonyms: RAC1. Accession Number: P63000. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.