Learn More
Biomatik Corporation GTPBP6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09045
Encompass_Preferred
The GTPBP6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Putative GTP-binding protein 6 protein (146-295AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Putative GTP-binding protein 6, GTPBP6, PGPL, Pseudoautosomal GTP-binding protein-like. UniProt: O43824. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8628