Learn More
Biomatik Corporation LYPLA2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508641
Encompass_Preferred
Lypla2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LYPLA2. The Lypla2 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Acyl-Protein Thioesterase 2 Protein (1-226Aa). Applications: ELISA, IF. Target Synonyms: LYPLA2. Accession Number: O95372. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8641 Supplier Biomatik Corporation Supplier No. CAC09058
Lypla2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LYPLA2. The Lypla2 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Acyl-Protein Thioesterase 2 Protein (1-226Aa). Applications: ELISA, IF. Target Synonyms: LYPLA2. Accession Number: O95372. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.