Learn More
Biomatik Corporation CCL4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09072
Encompass_Preferred
The CCL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human C-C motif chemokine 4 protein (24-84AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: C-C motif chemokine 4, G-26 T-lymphocyte-secreted protein, HC21, Lymphocyte activation gene 1 protein, LAG-1, MIP-1-beta (1-69), Macrophage infl, CCL4, LAG1, MIP1B, SCYA4. UniProt: P13236. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8655