Learn More
Biomatik Corporation ACSL4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508658
Encompass_Preferred
The ACSL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Long-chain-fatty-acid--CoA ligase 4 protein (457-670AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Long-chain-fatty-acid--CoA ligase 4, Long-chain acyl-CoA synthetase 4, LACS 4, ACSL4, ACS4, FACL4, LACS4. UniProt: O60488. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8658 Supplier Biomatik Corporation Supplier No. CAC09075
The ACSL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Long-chain-fatty-acid--CoA ligase 4 protein (457-670AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Long-chain-fatty-acid--CoA ligase 4, Long-chain acyl-CoA synthetase 4, LACS 4, ACSL4, ACS4, FACL4, LACS4. UniProt: O60488. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.