Learn More
Biomatik Corporation DKK1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508660
Encompass_Preferred
The DKK1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Dickkopf-related protein 1 protein (33-263AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Dickkopf-related protein 1, Dickkopf-1, Dkk-1, hDkk-1, SK, DKK1, UNQ492/PRO1008. UniProt: O94907. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8660 Supplier Biomatik Corporation Supplier No. CAC09077
The DKK1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Dickkopf-related protein 1 protein (33-263AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Dickkopf-related protein 1, Dickkopf-1, Dkk-1, hDkk-1, SK, DKK1, UNQ492/PRO1008. UniProt: O94907. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.