Learn More
Biomatik Corporation CSF2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508664
Encompass_Preferred
The CSF2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Granulocyte-macrophage colony-stimulating factor protein (18-144AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: GM-CSF, Colony-stimulating factor, CSF, Molgramostin, Sargramostim, CSF2, GMCSF. UniProt: P04141. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8664 Supplier Biomatik Corporation Supplier No. CAC09081
The CSF2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Granulocyte-macrophage colony-stimulating factor protein (18-144AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: GM-CSF, Colony-stimulating factor, CSF, Molgramostin, Sargramostim, CSF2, GMCSF. UniProt: P04141. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.