Learn More
Biomatik Corporation IL17A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09083
Encompass_Preferred
Il17A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL17A. The Il17A Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-17A Protein (26-132Aa). Applications: ELISA, IHC, IF. Target Synonyms: IL17A. Accession Number: Q16552. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8666