Learn More
Biomatik Corporation LCN2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508668
Encompass_Preferred
Lcn2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LCN2. The Lcn2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ngal Protein (1-198Aa). Applications: ELISA, WB. Target Synonyms: LCN2. Accession Number: P80188. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8668 Supplier Biomatik Corporation Supplier No. CAC09085
Lcn2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LCN2. The Lcn2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ngal Protein (1-198Aa). Applications: ELISA, WB. Target Synonyms: LCN2. Accession Number: P80188. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.