Learn More
Biomatik Corporation SPP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508672
Encompass_Preferred
Spp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SPP1. The Spp1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Osteopontin Protein (17-302Aa). Applications: ELISA, IHC. Target Synonyms: SPP1. Accession Number: P10451. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8672 Supplier Biomatik Corporation Supplier No. CAC09089
Spp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SPP1. The Spp1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Osteopontin Protein (17-302Aa). Applications: ELISA, IHC. Target Synonyms: SPP1. Accession Number: P10451. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.