Learn More
Biomatik Corporation PAPPA anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09090
Encompass_Preferred
Pappa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PAPPA. The Pappa Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Pappalysin-1 Protein (95-302Aa). Applications: ELISA, IHC. Target Synonyms: PAPPA. Accession Number: Q13219. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8673