Learn More
Biomatik Corporation FPR1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09092
Encompass_Preferred
The FPR1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human fMet-Leu-Phe receptor protein (306-350AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: fMet-Leu-Phe receptor, fMLP receptor, FPR1, N-formyl peptide receptor, FPR, N-formylpeptide chemoattractant receptor. UniProt: P21462. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8675