Learn More
Biomatik Corporation NPPB anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09098
Encompass_Preferred
Nppb Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NPPB. The Nppb Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Natriuretic Peptides B Protein (27-102Aa). Applications: ELISA, WB. Target Synonyms: NPPB. Accession Number: P16860. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8681