Learn More
Biomatik Corporation L1 anti-Human papillomavirus type 18, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09105
Encompass_Preferred
L1 Polyclonal Antibody is a Human (Homo sapiens) papillomavirus type 18 Polyclonal Antibody to L1. The L1 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens) papillomavirus type 18. Immunogen: Recombinant Hpv18 Major Capsid Protein L1 Protein (1-568Aa). Applications: ELISA, WB. Target Synonyms: L1. Accession Number: P06794. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8688