Learn More
Biomatik Corporation CD36 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09109
Encompass_Preferred
The CD36 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Platelet glycoprotein 4 protein (191-376AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Platelet glycoprotein 4, Fatty acid translocase, FAT, Glycoprotein IIIb, GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, Platelet glycoprotein IV, GPIV, Thrombospondin receptor, CD36, CD36, GP3B, GP4. UniProt: P16671. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8692