Learn More
Biomatik Corporation ERBB2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09113
Encompass_Preferred
The ERBB2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Receptor tyrosine-protein kinase erbB-2 protein (153-598AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Metastatic lymph node gene 19 protein, Proto-oncogene Neu, Proto-oncogene c-ErbB-2, Tyrosine kinase-type cell surface receptor HER2, p185erbB2, CD_antigen, ERBB2, HER2, MLN19, NEU, NGL. UniProt: P04626. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8696