Learn More
Biomatik Corporation PTH anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508697
Encompass_Preferred
Pth Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTH. The Pth Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Parathyroid Hormone Protein (32-115Aa). Applications: ELISA, IHC. Target Synonyms: PTH. Accession Number: P01270. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8697 Supplier Biomatik Corporation Supplier No. CAC09114
Pth Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTH. The Pth Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Parathyroid Hormone Protein (32-115Aa). Applications: ELISA, IHC. Target Synonyms: PTH. Accession Number: P01270. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.