Learn More
Biomatik Corporation EGFR anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09124
Encompass_Preferred
The EGFR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Epidermal growth factor receptor protein (26-645AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Epidermal growth factor receptor 5, CLEC14A, C14orf27, EGFR5, UNQ236/PRO269. UniProt: P00533. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8707