Learn More
Biomatik Corporation CD3E anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09127
Encompass_Preferred
The CD3E Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human T-cell surface glycoprotein CD3 epsilon chain protein (23-207AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. UniProt: P07766. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8710