Learn More
Biomatik Corporation MDK anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09128
Encompass_Preferred
Mdk Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MDK. The Mdk Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Midkine Protein (21-143Aa). Applications: ELISA, IHC, IF. Target Synonyms: MDK. Accession Number: P21741. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8711