Learn More
Biomatik Corporation C3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508712
Encompass_Preferred
The C3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complement C3 protein (26-225AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3, CPAMD1. UniProt: P01024. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8712 Supplier Biomatik Corporation Supplier No. CAC09129
The C3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complement C3 protein (26-225AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3, CPAMD1. UniProt: P01024. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.