Learn More
Biomatik Corporation BMP6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09130
Encompass_Preferred
The BMP6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Bone morphogenetic protein 6 protein (382-513AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: BMP6, VGR. UniProt: P22004. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8713