Learn More
Biomatik Corporation AVP anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09136
Encompass_Preferred
The AVP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Vasopressin-neurophysin 2-copeptin protein (126-164AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:200-1000, IHC:1:20-1:200. Alternative Names: AVP-NPII, AVP, ARVP, VP. UniProt: P01185. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8719