Learn More
Biomatik Corporation AREG anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09140
Encompass_Preferred
The AREG Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Amphiregulin protein (107-148AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Amphiregulin1 Publication, Colorectum cell-derived growth factor1, AREG. UniProt: P15514. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8723