Learn More
Biomatik Corporation FTL anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508725
Encompass_Preferred
The FTL Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Ferritin light chain protein (2-175AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: FTL, Ferritin L subunit. UniProt: P02792. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8725 Supplier Biomatik Corporation Supplier No. CAC09142
The FTL Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Ferritin light chain protein (2-175AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: FTL, Ferritin L subunit. UniProt: P02792. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.