Learn More
Biomatik Corporation BGLAP anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508735
Encompass_Preferred
The BGLAP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Osteocalcin protein (52-100AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, BGLAP. UniProt: P02818. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8735 Supplier Biomatik Corporation Supplier No. CAC09152
The BGLAP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Osteocalcin protein (52-100AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, BGLAP. UniProt: P02818. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.