Learn More
Biomatik Corporation INS anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09153
Encompass_Preferred
Ins Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to INS. The Ins Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Insulin Protein (25-110Aa). Applications: ELISA, IHC. Target Synonyms: INS. Accession Number: P01308. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8736