Learn More
Biomatik Corporation IL17RA anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09161
Encompass_Preferred
Il17Ra Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL17RA. The Il17Ra Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-17 Receptor A Protein (552-743Aa). Applications: ELISA, WB, IF. Target Synonyms: IL17RA. Accession Number: Q96F46. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8744