Learn More
Biomatik Corporation BCL2 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09162
Encompass_Preferred
The BCL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Apoptosis regulator Bcl-2 protein (31-80AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:200-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Apoptosis regulator Bcl-2, BCL2. UniProt: P10415. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8745