Learn More
Biomatik Corporation APX1 anti-Arabidopsis thaliana, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09166
Encompass_Preferred
The APX1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Arabidopsis thaliana. Immunogen: Recombinant Arabidopsis thaliana APX1 protein (3-247AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:2000. Alternative Names: APX1. UniProt: Q05431. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8749